siRNA Details
| Gene Symbol: St5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001326.1 | NP_001001326.1 | 4 | 1532 | ||
| NM_029811.2 | NP_084087.2 | 3 | 281 | |||
| GenBank mRNA | AK158270.1 | - | 4 | 1537 | ||
| BC051034.1 | - | 4 | 1713 | 5345299 | ||
| CF737707.1 | - | 1 | 14 | |||
| KC789645.1 | - | 4 | 1132 | |||

St5
Genomic Map