siRNA Details
| Gene Symbol: 2700049A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163378.1 | NP_001156850.1 | 14 | 3041 | ||
| NM_029818.1 | NP_084094.1 | 14 | 3041 | |||
| GenBank mRNA | AK172976.1 | - | 14 | 2691 | ||
| BC043104.1 | - | 14 | 3041 | |||
| BC150744.1 | - | 14 | 2409 | 9087655 | ||
 Back To Top
 Back To Top
 2700049A03Rik
2700049A03Rik Genomic Map
Genomic Map Back To Top
 Back To Top