siRNA Details
| Gene Symbol: Urah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029821.2 | NP_084097.1 | 2 | 295 | ||
| GenBank mRNA | AK004470.1 | - | 2 | 513 | ||
| AK013117.1 | - | 2 | 297 | |||
| BC051545.1 | - | 2 | 463 | 5255010 | ||
| DQ225768.1 | - | 2 | 170 | |||

Urah
Genomic Map