siRNA Details
| Gene Symbol: Arfgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166024.1 | NP_001159496.1 | 6, 7 | 771 | ||
| NM_023854.2 | NP_076343.2 | 6, 7 | 771 | |||
| GenBank mRNA | AF229439.1 | - | 6, 7 | 580 | ||
| AK009565.1 | - | 6, 7 | 595 | |||
| AK149837.1 | - | 6, 7 | 606 | |||
| AK168489.1 | - | 6, 7 | 581 | |||
| AK169144.1 | - | 6, 7 | 594 | |||
| BC005495.1 | - | 6, 7 | 619 | 3487811 | ||

Arfgap2
Genomic Map