siRNA Details
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205391.1 | NP_001192320.1 | 13 | 1416 | ||
NM_001205392.1 | NP_001192321.1 | 11 | 1311 | |||
NM_029846.4 | NP_084122.2 | 12 | 1368 | |||
GenBank mRNA | AB087879.1 | - | 11 | 1122 | ||
AB087880.1 | - | 12 | 1179 | |||
AB087881.1 | - | 13 | 1227 | |||
AB476646.1 | - | 13 | 1227 | |||
AK005181.1 | - | 13 | 1341 | |||
AK030983.1 | - | 12 | 1309 | |||
AK131120.1 | - | 11 | 2154 | |||
AK155358.1 | - | 11 | 1247 | |||
AK163246.1 | - | 11 | 1250 | |||
AK170073.1 | - | 11 | 1245 | |||
BC033362.1 | - | 3 | 689 | |||
BC049122.1 | - | 13 | 1341 | 6813377 | ||
BC052087.1 | - | 1 | 22 |
