siRNA Details
| Gene Symbol: Ccdc183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029859.1 | NP_084135.1 | 10, 11 | 1180 | ||
| GenBank mRNA | AK014983.1 | - | 8, 9 | 2012 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc183| Gene Symbol: Ccdc183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029859.1 | NP_084135.1 | 10, 11 | 1180 | ||
| GenBank mRNA | AK014983.1 | - | 8, 9 | 2012 | ||