siRNA Details
Gene Symbol: Ccdc183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029859.1 | NP_084135.1 | 11 | 1303 | ||
GenBank mRNA | AK014983.1 | - | 9 | 2135 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029859.1 | NP_084135.1 | 11 | 1303 | ||
GenBank mRNA | AK014983.1 | - | 9 | 2135 |