siRNA Details
| Gene Symbol: Ints7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034856.1 | - | 15 | 2158 | ||
| AK052103.1 | - | 13 | 1886 | |||
| AK085336.1 | - | 14 | 2033 | |||
| AK086462.1 | - | 15 | 2190 | |||
| BC054120.1 | - | 15 | 2143 | 6506658 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ints7| Gene Symbol: Ints7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034856.1 | - | 15 | 2158 | ||
| AK052103.1 | - | 13 | 1886 | |||
| AK085336.1 | - | 14 | 2033 | |||
| AK086462.1 | - | 15 | 2190 | |||
| BC054120.1 | - | 15 | 2143 | 6506658 | ||