siRNA Details
Gene Symbol: Tmem181b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039520.1 | - | 4 | 542 | ||
AK137412.1 | - | 5 | 1916 | |||
BC032285.1 | - | 4 | 1797 | |||
BC147312.1 | - | 1122 | 9056375 | |||
BC147313.1 | - | 1122 | 9056366 |
Gene Symbol: Tmem181a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033178.3 | NP_001028350.1 | 12 | 1142 | ||
GenBank mRNA | AK122507.1 | - | 9 | 2410 | ||
AK154554.1 | - | 12 | 1138 | |||
BE862876.1 | - | 4 | 315 |
