siRNA Details
| Gene Symbol: 8030423J24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029873.1 | NP_084149.1 | 3 | 407 | ||
| GenBank mRNA | AK020194.1 | - | 3 | 408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8030423J24Rik| Gene Symbol: 8030423J24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029873.1 | NP_084149.1 | 3 | 407 | ||
| GenBank mRNA | AK020194.1 | - | 3 | 408 | ||