siRNA Details
| Gene Symbol: Ptgr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252625.1 | NP_001239554.1 | 9 | 1242 | ||
| NM_001252626.1 | NP_001239555.1 | 10 | 1314 | |||
| NM_029880.3 | NP_084156.2 | 9 | 1185 | |||
| GenBank mRNA | AK007522.1 | - | 10 | 1198 | ||
| AK020666.1 | - | 9 | 1119 | |||
| AK021033.1 | - | 9 | 1077 | |||
| AK159187.1 | - | 9 | 1097 | |||
| AK159932.1 | - | 9 | 1082 | |||
| AK168895.1 | - | 9 | 1076 | |||

Ptgr2
Genomic Map