siRNA Details
| Gene Symbol: Fam162b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029894.1 | NP_084170.1 | 2, 3 | 314 | ||
| GenBank mRNA | AK020485.1 | - | 2, 3 | 335 | ||
| BC147085.1 | - | 2, 3 | 314 | 9056123 | ||
| BC147086.1 | - | 2, 3 | 314 | 9056133 | ||

Fam162b
Genomic Map