siRNA Details
| Gene Symbol: Stk31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029916.2 | NP_084192.2 | 9 | 1127 | ||
| GenBank mRNA | AF285580.1 | - | 9 | 1127 | ||
| BC137989.1 | - | 9 | 1124 | 8861008 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk31| Gene Symbol: Stk31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029916.2 | NP_084192.2 | 9 | 1127 | ||
| GenBank mRNA | AF285580.1 | - | 9 | 1127 | ||
| BC137989.1 | - | 9 | 1124 | 8861008 | ||