siRNA Details
| Gene Symbol: Anks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128086.2 | NP_001121558.2 | 13 | 2294 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anks1b| Gene Symbol: Anks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128086.2 | NP_001121558.2 | 13 | 2294 | ||