siRNA Details
| Gene Symbol: Shisa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175259.4 | NP_780468.1 | 5 | 676 | ||
| GenBank mRNA | AK039457.1 | - | 5 | 687 | ||
| AK134719.1 | - | 3 | 459 | |||
| BC044813.1 | - | 5 | 676 | 4950177 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shisa4| Gene Symbol: Shisa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175259.4 | NP_780468.1 | 5 | 676 | ||
| GenBank mRNA | AK039457.1 | - | 5 | 687 | ||
| AK134719.1 | - | 3 | 459 | |||
| BC044813.1 | - | 5 | 676 | 4950177 | ||