siRNA Details
| Gene Symbol: H2afv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029938.1 | NP_084214.1 | 4 | 390 | ||
| GenBank mRNA | AK021267.1 | - | 4 | 313 | ||
| AK155105.1 | - | 4 | 393 | |||
| AK168110.1 | - | 4 | 362 | |||
| BC028539.1 | - | 3 | 185 | |||
| BC147293.1 | - | 3 | 349 | 9056353 | ||
| BC147294.1 | - | 4 | 375 | 9056344 | ||
| BC147741.1 | - | 4 | 375 | 9056690 | ||
| BC147751.1 | - | 4 | 375 | 9056700 | ||
| CJ069840.1 | - | 4 | 391 | |||

H2afv
Genomic Map