siRNA Details
| Gene Symbol: H2afv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029938.1 | NP_084214.1 | 5 | 456 | ||
| GenBank mRNA | AK021267.1 | - | 5 | 379 | ||
| AK155105.1 | - | 5 | 459 | |||
| AK168110.1 | - | 5 | 428 | |||
| BC028539.1 | - | 4 | 251 | |||
| BC147293.1 | - | 4 | 415 | 9056353 | ||
| BC147294.1 | - | 5 | 441 | 9056344 | ||
| BC147741.1 | - | 5 | 441 | 9056690 | ||
| BC147751.1 | - | 5 | 441 | 9056700 | ||

H2afv
Genomic Map