siRNA Details
| Gene Symbol: Ehmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012518.3 | NP_001012536.2 | 6 | 1125 | ||
| NM_001109686.2 | NP_001103156.1 | 6 | 1104 | |||
| NM_001109687.2 | NP_001103157.1 | 6 | 1125 | |||
| NM_172545.4 | NP_766133.2 | 6 | 1104 | |||
| GenBank mRNA | AB205007.1 | - | 6 | 1131 | ||
| AK049454.1 | - | 3 | 375 | |||
| AK052174.1 | - | 6 | 1088 | |||
| AK082062.1 | - | 6 | 1115 | |||
| BC056938.1 | - | 4 | 826 | |||
| BC089302.1 | - | 4 | 972 | |||

Ehmt1
Genomic Map