siRNA Details
| Gene Symbol: Ehmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012518.3 | NP_001012536.2 | 6 | 1034 | ||
| NM_001109686.2 | NP_001103156.1 | 6 | 1013 | |||
| NM_001109687.2 | NP_001103157.1 | 6 | 1034 | |||
| NM_172545.4 | NP_766133.2 | 6 | 1013 | |||
| GenBank mRNA | AB205007.1 | - | 6 | 1040 | ||
| AK049454.1 | - | 3 | 284 | |||
| AK052174.1 | - | 6 | 996 | |||
| AK054555.1 | - | 6 | 981 | |||
| AK082062.1 | - | 6 | 1024 | |||
| BC056938.1 | - | 4 | 735 | |||
| BC089302.1 | - | 4 | 881 | |||

Ehmt1
Genomic Map