siRNA Details
| Gene Symbol: 6030468B19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029964.1 | NP_084240.1 | 3 | 231 | ||
| GenBank mRNA | AK020081.1 | - | 3 | 156 | ||
| BB843270.1 | - | 3 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6030468B19Rik| Gene Symbol: 6030468B19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029964.1 | NP_084240.1 | 3 | 231 | ||
| GenBank mRNA | AK020081.1 | - | 3 | 156 | ||
| BB843270.1 | - | 3 | 232 | |||