siRNA Details
| Gene Symbol: Rnf170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029965.2 | NP_084241.1 | 8 | 1036 | ||
| GenBank mRNA | AK036051.1 | - | 8 | 895 | ||
| AK042063.1 | - | 7 | 892 | |||
| AK153658.1 | - | 7 | 963 | |||
| AK162554.1 | - | 8 | 1051 | |||
| BC031383.1 | - | 8 | 821 | |||
| BC114210.1 | - | 8 | 1036 | 6309632 | ||
| BC138930.1 | - | 8 | 864 | 8861952 | ||
| BC145718.1 | - | 8 | 869 | 40131021 | ||

Rnf170
Genomic Map