siRNA Details
| Gene Symbol: Elp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023876.3 | NP_076365.2 | 5 | 633 | ||
| GenBank mRNA | AJ276004.1 | - | 5 | 632 | ||
| AK020742.1 | - | 5 | 641 | |||
| AK034040.1 | - | 5 | 638 | |||
| AK034529.1 | - | 5 | 639 | |||
| AK037104.1 | - | 5 | 657 | |||
| AK039447.1 | - | 5 | 645 | |||
| AK039758.1 | - | 5 | 634 | |||
| AK079069.1 | - | 5 | 648 | |||
| AK168964.1 | - | 5 | 697 | |||

Elp4
Genomic Map