siRNA Details
| Gene Symbol: Adamtsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029981.1 | NP_084257.1 | 7, 8 | 866 | ||
| GenBank mRNA | AB096928.1 | - | 7, 8 | 866 | ||
| AK156991.1 | - | 7, 8 | 869 | |||
| BC128568.1 | - | 7, 8 | 818 | 40097415 | ||
| BC129984.1 | - | 7, 8 | 818 | 40097418 | ||

Adamtsl2
Genomic Map