siRNA Details
| Gene Symbol: A930009A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029982.1 | NP_084258.1 | 4 | 439 | ||
| GenBank mRNA | AK020834.1 | - | 4 | 440 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930009A15Rik| Gene Symbol: A930009A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029982.1 | NP_084258.1 | 4 | 439 | ||
| GenBank mRNA | AK020834.1 | - | 4 | 440 | ||