siRNA Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133922.3 | NP_598683.3 | 13 | 2101 | ||
| GenBank mRNA | AB041543.1 | - | 11 | 1733 | ||
| AK029235.1 | - | 4 | 547 | |||
| AK040525.1 | - | 13 | 2103 | |||
| AK128931.1 | - | 13 | 2052 | |||
| AK162934.1 | - | 13 | 1809 | |||
| AK173281.1 | - | 13 | 3805 | |||
| BC066816.1 | - | 13 | 2021 | 30084911 | ||

Krba1
Genomic Map