siRNA Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133922.3 | NP_598683.3 | 15 | 2444 | ||
| GenBank mRNA | AB041543.1 | - | 13 | 2076 | ||
| AK029235.1 | - | 6 | 890 | |||
| AK038090.1 | - | 1 | 94 | |||
| AK040525.1 | - | 15 | 2446 | |||
| AK128931.1 | - | 15 | 2395 | |||
| AK162934.1 | - | 15 | 2152 | |||
| AK173281.1 | - | 15 | 4148 | |||
| AK196294.1 | - | 2 | 233 | |||
| BC066816.1 | - | 15 | 2364 | 30084911 | ||

Krba1
Genomic Map