siRNA Details
| Gene Symbol: Tchp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029992.2 | NP_084268.2 | 2 | 286 | ||
| GenBank mRNA | AK020912.1 | - | 2 | 249 | ||
| AK083027.1 | - | 2 | 202 | |||
| AK159947.1 | - | 2 | 247 | |||
| BC043048.1 | - | 2 | 191 | 5704036 | ||
| BU512279.1 | - | 2 | 286 | |||

Tchp
Genomic Map