siRNA Details
| Gene Symbol: Tchp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029992.2 | NP_084268.2 | 3 | 440 | ||
| GenBank mRNA | AK020912.1 | - | 3 | 403 | ||
| AK083027.1 | - | 3 | 356 | |||
| AK159947.1 | - | 3 | 401 | |||
| AK216033.1 | - | 2 | 158 | |||
| BC043048.1 | - | 3 | 345 | 5704036 | ||
| BU512279.1 | - | 3 | 440 | |||

Tchp
Genomic Map