siRNA Details
| Gene Symbol: Grifin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030022.1 | NP_084298.1 | 3 | 143 | ||
| GenBank mRNA | AK021365.1 | - | 3 | 144 | ||
| BC107323.2 | - | 3 | 143 | 40053455 | ||
| BC107324.1 | - | 3 | 143 | 40053456 | ||
| BC107343.2 | - | 3 | 401 | |||

Grifin
Genomic Map