siRNA Details
| Gene Symbol: Prr15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030024.2 | NP_084300.1 | 2 | 634 | ||
| GenBank mRNA | AJ132433.2 | - | 2 | 634 | ||
| AK021401.1 | - | 2 | 742 | |||
| AK136438.1 | - | 2 | 814 | |||
| BC055838.1 | - | 2 | 763 | |||
| BC132416.1 | - | 1 | 310 | 40130693 | ||
| BC138757.1 | - | 1 | 310 | 8861779 | ||

Prr15
Genomic Map