siRNA Details
| Gene Symbol: Tmem190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030028.1 | NP_084304.1 | 5 | 402 | ||
| GenBank mRNA | AK019807.1 | - | 5 | 395 | ||
| BU936271.1 | - | 405 | ||||
| BX631095.1 | - | 1 | 184 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem190| Gene Symbol: Tmem190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030028.1 | NP_084304.1 | 5 | 402 | ||
| GenBank mRNA | AK019807.1 | - | 5 | 395 | ||
| BU936271.1 | - | 405 | ||||
| BX631095.1 | - | 1 | 184 | |||