siRNA Details
| Gene Symbol: Cpt1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252470.1 | NP_001239399.1 | 6 | 925 | ||
| NM_153679.2 | NP_710146.1 | 6 | 927 | |||
| GenBank mRNA | AF320000.1 | - | 6 | 850 | ||
| AK035790.1 | - | 6 | 891 | |||
| AK087747.1 | - | 6 | 799 | |||
| BC066155.1 | - | 6 | 849 | 30089333 | ||
| BC083317.1 | - | 6 | 808 | |||

Cpt1c
Genomic Map