siRNA Details
| Gene Symbol: 9230117E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020351.1 | - | 2 | 176 | ||
| BY721164.1 | - | 2 | 176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9230117E06Rik| Gene Symbol: 9230117E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020351.1 | - | 2 | 176 | ||
| BY721164.1 | - | 2 | 176 | |||