siRNA Details
| Gene Symbol: 4930451I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183131.2 | NP_898954.2 | 2 | 195 | ||
| GenBank mRNA | AK019625.1 | - | 2 | 172 | ||
| BC048622.1 | - | 2 | 195 | 6743640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930451I11Rik| Gene Symbol: 4930451I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183131.2 | NP_898954.2 | 2 | 195 | ||
| GenBank mRNA | AK019625.1 | - | 2 | 172 | ||
| BC048622.1 | - | 2 | 195 | 6743640 | ||