siRNA Details
| Gene Symbol: Adgrf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289499.1 | NP_001276428.1 | 6 | 1650 | ||
| NM_001289500.1 | NP_001276429.1 | 6 | 1637 | |||
| NM_001289501.1 | NP_001276430.1 | 6 | 1681 | |||
| NM_030067.2 | NP_084343.1 | 6 | 1724 | |||
| GenBank mRNA | AK019508.1 | - | 6 | 1727 | ||
| BC089564.1 | - | 6 | 1650 | 30502915 | ||
| BC140980.1 | - | 6 | 1590 | 9055592 | ||
More Information
Other Species Targeted
Set Membership: |
|

Adgrf4
Genomic Map