siRNA Details
| Gene Symbol: Iqch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030068.1 | NP_084344.1 | 9, 10 | 891 | ||
| GenBank mRNA | AK019535.1 | - | 9, 10 | 923 | ||
| AK029502.1 | - | 9, 10 | 927 | |||
| BC141047.1 | - | 9, 10 | 891 | 9055659 | ||
| BC145231.1 | - | 9, 10 | 891 | 9053783 | ||

Iqch
Genomic Map