siRNA Details
| Gene Symbol: Zfp712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166218.1 | NP_001159690.1 | 4 | 1370 | ||
| GenBank mRNA | AK019536.1 | - | 4 | 1352 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp712| Gene Symbol: Zfp712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166218.1 | NP_001159690.1 | 4 | 1370 | ||
| GenBank mRNA | AK019536.1 | - | 4 | 1352 | ||