siRNA Details
| Gene Symbol: Mroh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030071.1 | NP_084347.1 | 7, 8 | 629 | ||
| GenBank mRNA | AK019544.1 | - | 7, 8 | 630 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mroh9| Gene Symbol: Mroh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030071.1 | NP_084347.1 | 7, 8 | 629 | ||
| GenBank mRNA | AK019544.1 | - | 7, 8 | 630 | ||