siRNA Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF376726.1 | - | 12 | 1195 | ||
| AK019012.1 | - | 3 | 169 | |||
| AK047364.1 | - | 12 | 1171 | |||
| AK082797.1 | - | 12 | 1220 | |||
| AK159313.1 | - | 12 | 1240 | |||
| AK170948.1 | - | 11 | 1323 | |||
| AK171030.1 | - | 11 | 1323 | |||
| AK185933.1 | - | 3 | 156 | |||
| AX775897.1 | - | 12 | 1195 | |||

Gpr108
Genomic Map