siRNA Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081335.2 | NP_001074804.2 | 32 | 6541 | ||
| GenBank mRNA | AK051474.1 | - | 9 | 1769 | ||
| AK122351.1 | - | 29 | 5531 | |||
| AK147675.1 | - | 30 | 7131 | |||
| AK162968.1 | - | 11 | 2002 | |||

Cul9
Genomic Map