siRNA Details
| Gene Symbol: 2700046A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012381.1 | - | 1, 2 | 344 | ||
| AK020725.1 | - | 1, 2 | 416 | |||
| BC072581.1 | - | 1, 2 | 214 | |||
| CJ067024.1 | - | 1, 2 | 384 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2700046A07Rik| Gene Symbol: 2700046A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012381.1 | - | 1, 2 | 344 | ||
| AK020725.1 | - | 1, 2 | 416 | |||
| BC072581.1 | - | 1, 2 | 214 | |||
| CJ067024.1 | - | 1, 2 | 384 | |||