siRNA Details
Gene Symbol: 2700046A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012381.1 | - | 3 | 445 | ||
BC072581.1 | - | 3 | 315 | |||
CJ067024.1 | - | 3 | 485 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2700046A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012381.1 | - | 3 | 445 | ||
BC072581.1 | - | 3 | 315 | |||
CJ067024.1 | - | 3 | 485 |