siRNA Details
Gene Symbol: Helz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198298.1 | NP_938040.1 | 22 | 3115 | ||
GenBank mRNA | AB093209.1 | - | 23 | 3173 | ||
AK033094.1 | - | 6 | 770 | |||
AK044226.1 | - | 7 | 974 | |||
BC076626.1 | - | 22 | 3115 | 30640711 |
