siRNA Details
| Gene Symbol: 1700084C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033185.2 | NP_001028357.1 | 2 | 138 | ||
| GenBank mRNA | AK141447.1 | - | 2 | 139 | ||
| BC150846.1 | - | 1 | 63 | 9087757 | ||
| BC172001.1 | - | 1 | 63 | 9054287 | ||

1700084C01Rik
Genomic Map