siRNA Details
| Gene Symbol: Skap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033186.3 | NP_001028358.1 | 7, 8 | 679 | ||
| NM_001177898.1 | NP_001171369.1 | 7, 8 | 679 | |||
| NM_001177899.1 | NP_001171370.1 | 7, 8 | 679 | |||
| GenBank mRNA | AK137792.1 | - | 5, 6 | 485 | ||
| AK137942.1 | - | 7, 8 | 647 | |||
| AK138030.1 | - | 7, 8 | 662 | |||
| AK169825.1 | - | 7, 8 | 644 | |||
| BC171961.1 | - | 7, 8 | 608 | 9054247 | ||

Skap1
Genomic Map