siRNA Details
| Gene Symbol: Arhgap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030113.2 | NP_084389.2 | 18 | 1696 | ||
| GenBank mRNA | AF297029.1 | - | 18 | 1600 | ||
| AF297030.1 | - | 18 | 1659 | |||
| AY179965.1 | - | 18 | 1572 | |||
| AY179966.1 | - | 17 | 1506 | |||
| AY179967.1 | - | 18 | 1572 | |||
| BU151977.1 | - | 8 | 522 | |||

Arhgap10
Genomic Map