siRNA Details
| Gene Symbol: Htra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042615.2 | NP_001036080.1 | 6 | 1473 | ||
| NM_030127.3 | NP_084403.2 | 6 | 1473 | |||
| GenBank mRNA | AK008764.1 | - | 5 | 595 | ||
| AK035194.1 | - | 6 | 1476 | |||
| AY037300.1 | - | 6 | 1258 | |||
| AY156509.1 | - | 6 | 1165 | |||
| AY280664.1 | - | 6 | 1165 | |||
| BC022646.1 | - | 5 | 584 | |||
| BC138587.1 | - | 6 | 1197 | 8861607 | ||
| BC138588.1 | - | 6 | 1197 | 8861608 | ||

Htra3
Genomic Map