siRNA Details
Gene Symbol: Utp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030132.5 | NP_084408.1 | 2 | 298 | ||
GenBank mRNA | AK005831.1 | - | 2 | 99 | ||
AK021306.1 | - | 2 | 248 | |||
AK042518.1 | - | 2 | 255 | |||
AK134445.1 | - | 2 | 230 | |||
AK188127.1 | - | 2 | 66 | |||
BC046791.1 | - | 2 | 243 | 5717427 | ||
BC056961.1 | - | 2 | 243 | 5717427 | ||
CK334116.1 | - | 2 | 310 |
