siRNA Details
| Gene Symbol: Bola3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175277.4 | NP_780486.1 | 2 | 162 | ||
| GenBank mRNA | AK028102.1 | - | 2 | 163 | ||
| AK131950.1 | - | 171 | ||||
| AK160488.1 | - | 2 | 166 | |||
| AK213244.1 | - | 1 | 97 | |||
| BC049748.1 | - | 2 | 182 | |||
| BC116947.1 | - | 2 | 200 | 40126266 | ||
| BC116949.1 | - | 2 | 200 | 40126268 | ||

Bola3
Genomic Map