siRNA Details
| Gene Symbol: Plekhj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023900.2 | NP_076389.2 | 3, 4 | 680 | ||
| GenBank mRNA | AK020621.1 | - | 3, 4 | 349 | ||
| AK051734.1 | - | 3, 4 | 337 | |||
| AK150144.1 | - | 3, 4 | 295 | |||
| AK178993.1 | - | 1, 2 | 85 | |||
| AK219000.1 | - | 1, 2 | 17 | |||
| BC005565.1 | - | 3, 4 | 278 | 3966305 | ||

Plekhj1
Genomic Map